Learn More
Hach Company Potassium Chloride Standard Solution, 35.0 ppt Salinity, 53.0 mS/cm Conductivity, 500 mL
Catalog No. 501996591 Shop All Hach Company Products
Encompass
For calibrations/accuracy checks on conductivity/salinity meters.
Catalog No. 50-199-6591 Supplier Hach Company Supplier No. 2714349
For calibrations/accuracy checks on conductivity/salinity meters.