Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Salinity Test
SDPSCI_ED

Catalog No. S450911 Shop All LaMotte Company Products
Change view
Click to view available options
No. of Tests:
50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tests
S450911 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S450911 Supplier LaMotte™ Supplier No. 745902
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Test sea water, marine estuaries, aquariums and brine solutions.

  • Uses a direct-reading titrator
  • Conduct accurate salt water analysis
  • Measures 0 to 20ppt salinity in 0.4ppt increments
  • Range can be extended to 0 to 40ppt
  • Utilizes modified APHA argentometric method
  • Contains three reagents
  • Enough materials for 50 tests at 20ppt

Specifications

For Use With (Application) Environmental education
For Use With (Equipment) Direct reading titrator
Includes 3 reagents
No. of Tests 50
Range 0 to 40ppt
Type Salinity Water Test Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.