Learn More
Cayman Chemical Salinomycin, Purity: ≥90%, CAS: 53003-10-4, 10 mg
SDP
Catalog No. 502955182
Encompass_Preferred
An antibacterial and coccidiostat compound that shows selective toxicity for HMLER breast cancer stem cells (IC50 = ~24 μM); 8 μM treatment of 4T1 and MCF-7-Ras breast cancer cell lines decreases cancer stem cell growth ~2-fold and ~3-fold, respectively; 5 mg/kg inhibits mammary tumor growth and induces increased epithelial differentiation of tumor cells in mice implanted with SUM159 human breast cancer cells
Catalog No. 50-295-5182 Supplier Cayman Chemical Supplier No. 1357910 MG
An antibacterial and coccidiostat compound that shows selective toxicity for HMLER breast cancer stem cells (IC50 = ~24 μM); 8 μM treatment of 4T1 and MCF-7-Ras breast cancer cell lines decreases cancer stem cell growth ~2-fold and ~3-fold, respectively; 5 mg/kg inhibits mammary tumor growth and induces increased epithelial differentiation of tumor cells in mice implanted with SUM159 human breast cancer cells