Learn More
Apexbio Technology LLC Salinomycin, 5mg. Cas: 53003-10-4 MFCD: MFCD00865281. Polyether ionophore antibiotic;anti-cancer
SDP
Catalog No. 501904839
Encompass_Preferred
IC50: 7.7, 13.7 and 10.4 M for HepG2, SMMC-7721 and BEL-7402 cell line, respectively (after 24h treatment)Salinomycin (Sal), which is a polyether ionophore antibiotic from Streptomyces albus, has been proven to be able to kill different types of human cancer cells. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4839 Supplier Apexbio Technology LLC Supplier No. A37855
IC50: 7.7, 13.7 and 10.4 M for HepG2, SMMC-7721 and BEL-7402 cell line, respectively (after 24h treatment)Salinomycin (Sal), which is a polyether ionophore antibiotic from Streptomyces albus, has been proven to be able to kill different types of human cancer cells. Other sizes are available. Please inqury us for quote.