Learn More
Abcam Salinomycin sodium salt, cation ionophore, 50MG
Catalog No. 502704879
Encompass_Preferred
MW 772.98 g/mol. Cation ionophore. Antibiotic agent. Salinomycin (ab141186) salt. Shows cancer stems cell selective antiproliferative effects (IC₅₀ values are 24 and 90 μM for cancer stem cell subpopulation and HMLER breast cancer cells respectively). Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4879 Supplier Abcam Supplier No. AB14617850MG
MW 772.98 g/mol. Cation ionophore. Antibiotic agent. Salinomycin (ab141186) salt. Shows cancer stems cell selective antiproliferative effects (IC₅₀ values are 24 and 90 μM for cancer stem cell subpopulation and HMLER breast cancer cells respectively). Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement