Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salinomycin sodium salt, MP Biomedicals™

Catalog No. ICN15204350 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15204350 50 mg
ICN15204305 5 mg
ICN15204325 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15204350 Supplier MP Biomedicals, Inc Supplier No. 0215204350
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 55721-31-8
Molecular Formula C42H69NaO11
Molecular Weight (g/mol) 772.993
MDL Number MFCD00167109
InChI Key YPZYGIQXBGHDBH-IETZRCTKSA-M
Synonym Coxistal
PubChem CID 124080880
IUPAC Name sodium;(2S)-2-[(2R,5S,6S)-6-[(2R,3R,4R,6S)-6-[(3R,5S,7R,9S,10S,12R,15R)-3-[(2S,5R,6S)-5-ethyl-5-hydroxy-6-methyloxan-2-yl]-15-hydroxy-3,10,12-trimethyl-4,6,8-trioxadispiro[4.1.5^{7}.3^{5}]pentadec-13-en-9-yl]-3-hydroxy-4-methyl-5-oxooctan-2-yl]-5-methylox
SMILES CCC(C1CCC(C(O1)C(C)C(C(C)C(=O)C(CC)C2C(CC(C3(O2)C=CC(C4(O3)CCC(O4)(C)C5CCC(C(O5)C)(CC)O)O)C)C)O)C)C(=O)[O-].[Na+]

Specifications

Type Salinomycin Sodium Salt
Physical Form Powder
Color White
Quantity 50 mg
Formula Weight 772.98
Grade Not Available
Chemical Name or Material Salinomycin sodium salt, Molecular biology
EINECSNumber 258-290-1
Recommended Storage Store at 4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.