Learn More
Apexbio Technology LLC Salirasib 162520-00-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063942
Encompass_Preferred
Salirasib (CAS 162520-00-5) chemically known as S-trans trans-farnesylthiosalicylic acid (FTS) is a synthetic inhibitor targeting Ras proteins Structurally mimicking the carboxy-terminal farnesylated cysteine found in Ras Salirasib disrupts Ras localization at the cellular membrane thereby reducing activity of H-ras K-ras and N-ras isoforms Research indicates that Salirasib reduces Ras protein levels by approximately 50% at concentrations of 25 50 M in Panc-1 pancreatic cancer cells In xenograft models it exhibits tumor growth inhibition alone and synergistically with gemcitabine Clinical phase I studies support its continued investigation in solid-tumor therapy recommending a dose of 600 mg/day
Catalog No. 50-000-63942 Supplier Apexbio Technology LLC Supplier No. A37875.1
Salirasib (CAS 162520-00-5) chemically known as S-trans trans-farnesylthiosalicylic acid (FTS) is a synthetic inhibitor targeting Ras proteins Structurally mimicking the carboxy-terminal farnesylated cysteine found in Ras Salirasib disrupts Ras localization at the cellular membrane thereby reducing activity of H-ras K-ras and N-ras isoforms Research indicates that Salirasib reduces Ras protein levels by approximately 50% at concentrations of 25 50 M in Panc-1 pancreatic cancer cells In xenograft models it exhibits tumor growth inhibition alone and synergistically with gemcitabine Clinical phase I studies support its continued investigation in solid-tumor therapy recommending a dose of 600 mg/day