Learn More
Apexbio Technology LLC Salirasib,25mg CAS# 162520-00-5
SDP
Supplier:  Apexbio Technology LLC A378725
Encompass_Preferred
Salirasib (S-trans,trans-farnesylthiosalycilic acid [FTS]) is a synthetic small molecule that acts as a potent Ras inhibitor.[1]The Ras family of small GTPases transmits extracellular signals, which are initiated by cell-surface receptors and serve to regulate various cellular processes including cell growth, differentiation, motility and cell death. Signals transmitted by activated Ras induce activation of multiple effectors. Ras signaling is activated in a large number of human cancers. Mutations of codons 12, 13 and 61 in Ras result in constitutively active Ras, and activating mutations of the three major Ras isoforms (H, K and N) have been found in more than 33% of human cancers. [2] Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0219