Learn More
TARGETMOL CHEMICALS INC SALIRASIB 50MG
SDP
Catalog No. 502579509
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Salirasib (Farnesyl Thiosalicylic Acid)(Ki=2.6 uM) an effective competitive prenylated protein methyltransferase (PPMTase) inhibitor inhibits Ras methylation with potential antineoplastic activity. purity: 99%
Catalog No. 50-257-9509 Supplier TARGETMOL CHEMICALS INC Supplier No. T616350MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Salirasib (Farnesyl Thiosalicylic Acid)(Ki=2.6 uM) an effective competitive prenylated protein methyltransferase (PPMTase) inhibitor inhibits Ras methylation with potential antineoplastic activity. purity: 99%