Learn More
Medchemexpress LLC HY-14754 100mg Medchemexpress, Salirasib S-Farnesylthiosalicylic acid;Farnesyl Thiosalicylic Acid;FTS CAS:162520-00-5 Purity:98%
SDP
Catalog No. 501638186
Encompass_Preferred
Medchemexpress,HY-14754 100mg Salirasib S-Farnesylthiosalicylic acid;Farnesyl Thiosalicylic Acid;FTS CAS:162520-00-5 Formula:C22H30O2S Salirasib is a potent and competitive prenylated protein methyltransferase (PPMTase) inhibitor with Ki of 2.6 μM, which inhibits Ras methylation. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8186 Supplier Medchemexpress LLC Supplier No. HY14754100MG
Medchemexpress,HY-14754 100mg Salirasib S-Farnesylthiosalicylic acid;Farnesyl Thiosalicylic Acid;FTS CAS:162520-00-5 Formula:C22H30O2S Salirasib is a potent and competitive prenylated protein methyltransferase (PPMTase) inhibitor with Ki of 2.6 μM, which inhibits Ras methylation. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.