Learn More
Honeywell Salisbury™ 31 cal/cm2 Flash Kits without Gloves
Catalog No. 19138166F Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-138-166F 3X-Large
19-138-166E 2X-Large
19-138-166C Large
19-138-166A Small
19-138-166B Medium
19-138-166D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-138-166F Supplier Honeywell Salisbury™ Supplier No. SK313XL
This item is not returnable. View return policy
Specifications
Arc Flash Rating 31 cal/cm²
Description Includes: Coat, bib overalls, Pro-Hood™, hard hat, safety Glasses and storage bag
Certifications/Compliance ASTM F1506, NFPA 70E™
Size 3X-Large