Learn More
Honeywell Salisbury™ Arc Flash Coverall Kit
Catalog No. 19139212 Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Description:
42 to 44 in. Chest navy blue coveralls, face shield, hard hat, safety Glasses and Salisbury back pack
46 to 48 in. Chest navy blue coveralls, face shield, hard hat, safety Glasses and Salisbury back pack
50 to 52 in. Chest navy blue coveralls, face shield, hard hat, safety Glasses and Salisbury back pack
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Size
19-139-212 50 to 52 in. Chest navy blue coveralls, face shield, hard hat, safety Glasses and Salisbury back pack 2X-Large
19-139-210 42 to 44 in. Chest navy blue coveralls, face shield, hard hat, safety Glasses and Salisbury back pack Large
19-139-211 46 to 48 in. Chest navy blue coveralls, face shield, hard hat, safety Glasses and Salisbury back pack X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-139-212 Supplier Honeywell Salisbury™ Supplier No. SKCA112XLBP
This item is not returnable. View return policy
Kit contains arc flash coveralls, an AS1000HAT, an appropriate ATPV-rated AFHOOD, ASBAG, SKBAG and safety glasses
TRUSTED_SUSTAINABILITY
Specifications
Arc Flash Rating 12 cal/cm²
Description 50 to 52 in. Chest navy blue coveralls, face shield, hard hat, safety Glasses and Salisbury back pack
Size 2X-Large
Certifications/Compliance NFPA 70E™ 2009, CE 0120
Color Navy
Material Cotton