Learn More
Honeywell Salisbury™ Arc Flash Coveralls
Catalog No. 18150202 Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-150-202 X-Large
18-150-201 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-150-202 Supplier Honeywell Salisbury™ Supplier No. ACCA8BLXL
This item is not returnable. View return policy
Specifications
Size X-Large
Material Cotton
Color Navy
Arc Flash Rating 8 Calories/cm2
Certifications/Compliance ASTM F1506, NFPA 70E™, CE 0120
Size (English) Chest 42 to 44 in.
Size (English) Inseam 30 in.
Size (Metric) Inseam 76.2 cm