Learn More
MSC Salisbury by Honeywell Leather Protectors, Cowhide, Large, 10 inch
Catalog No. 5000508099
Encompass
These leather protectors are designed for various applications, providing hand protection. Made from durable cowhide leather, they are size large with a 10-inch length, and are sold as a pair.
  • Continuous and rolled cuff
  • Sold as a pair
  • Not to be used for electrical protection according to ASTM F696-02 standards
  • Complies with ASTM F496 standards
Catalog No. 50-005-08099 Supplier MSC Supplier No. 00041038
These leather protectors are designed for various applications, providing hand protection. Made from durable cowhide leather, they are size large with a 10-inch length, and are sold as a pair.
  • Continuous and rolled cuff
  • Sold as a pair
  • Not to be used for electrical protection according to ASTM F696-02 standards
  • Complies with ASTM F496 standards