Learn More
Honeywell Salisbury™ G99 Portable Glove Inflator Kit - Replacement Parts
Catalog No. 19138421 Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Product Type:
Check Valve
Catalog No. Product Type
19-138-421 Check Valve
1 options
This item is not returnable. View return policy
Catalog No. 19-138-421 Supplier Honeywell Salisbury™ Supplier No. G99V
This item is not returnable. View return policy
The G99 Inflator Kit provides a quality means of inspecting gloves in the field. Replacement parts ensure accurate operation and help to extend product lifetime.
  • The G99 is a simple, easy to use, portable glove inflator
  • Provides a quality means of inspecting gloves in the field
  • Glove is secured to the inflator by a nylon strap, and fastened with a hook and pile closure
  • Inflation is accomplished by pumping the bellows of the inflator against any surface
  • Type I gloves should be inflated no more than 1.5x their normal size
  • Type II Salcor™ gloves should be inflated no more than 1.25x their normal size
  • Replacement parts help ensure that the pump is working properly whenever and wherever it is needed
TRUSTED_SUSTAINABILITY
Specifications
Product Type Check Valve
For Use With (Equipment) G99 Portable Glove Inflator Kit