Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Salisbury™ Insulated Rescue Hook

Catalog No. p-2715223 Shop All WH Salisbury & Co. Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Length (English):
96 in.
Length (Metric):
240 cm
Weight (English):
5.5 lb.
Weight (Metric):
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Length (Metric) Weight (English) Weight (Metric)
19-138-496 96 in. 240 cm 5.5 lb. 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-138-496 Supplier Honeywell Salisbury™ Supplier No. 24403
Only null left
Add to Cart
Add to Cart

Used to withdraw injured workers from hazardous locations

  • For use in confined spaces, in vaults, or near electrical cabinets and switch gear
  • Foam-filled, fiberglass reinforced handle
  • Coated, heat-treated body hook with 18 in. opening
  • Available in 6 or 8 ft. (1.8 or 2.4m) lengths
TRUSTED_SUSTAINABILITY

Specifications

Product Type Rescue Hook
Material Foam-filled Fiberglass
For Use With (Application) Confined spaces, in vaults or near electrical cabinets and switch gear
Includes Coated, heat treated body hook with 18 in. opening
Length (English) 96 in.
Length (Metric) 240 cm
Weight (English) 5.5 lb.
Weight (Metric) 2.5 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.