Learn More
MSC Salisbury Leather Protector Gloves, Goatskin, 10 in., 1 Pair
Catalog No. 5000769712
Encompass
Salisbury ILPG10 leather protector gloves are designed to be worn over insulating rubber gloves, providing mechanical protection against cuts, abrasions, and punctures. They are manufactured from top grain goatskin.
  • Provides mechanical protection against cuts, abrasions, and punctures
  • Manufactured from top grain goatskin
  • Overall length of 10 in.
  • Available in dual sizes 7/7.5 through 11/11.5 and 12
  • Compatible with Class 0 and 00 rubber gloves per ASTM F496 specifications
Catalog No. 50-007-69712 Supplier MSC Supplier No. 42244780
Salisbury ILPG10 leather protector gloves are designed to be worn over insulating rubber gloves, providing mechanical protection against cuts, abrasions, and punctures. They are manufactured from top grain goatskin.
  • Provides mechanical protection against cuts, abrasions, and punctures
  • Manufactured from top grain goatskin
  • Overall length of 10 in.
  • Available in dual sizes 7/7.5 through 11/11.5 and 12
  • Compatible with Class 0 and 00 rubber gloves per ASTM F496 specifications