Learn More
Honeywell Salisbury™ PRO-HOOD™ Arc Flash Protective Hoods
Catalog No. 19138110 Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Size:
Universal
Catalog No. Size
19-138-110 Universal
1 options
This item is not returnable. View return policy
Catalog No. 19-138-110 Supplier Honeywell Salisbury™ Supplier No. FH11BL
This item is not returnable. View return policy
Lighter tint lenses for better visual light transmission
  • 11 cal/cm2 to 100 cal/cm2 ATPV ratings
  • Made of arc-flash-resistant Indura™ Ultra Soft™ material and sewn with Nomex™ thread
  • 100 cal/cm2 is made from material layers of TuffWeld™ and Q/9
  • Arc-rated 10 × 20 in. antifogging replaceable lens
  • Slotted bracket
  • Hook-and-pile tabs to attach PRO-AIR belt-mounted compact air system
  • One size fits all
Compliance
Meet current ASTM™ F2178 and NFPA 70E™ standards and the flammability requirements for ASTM F1506.
†NFPA 70E does not have a Hazard Risk Category above 40 cal/cm2. Working on levels above 40cal/cm2 should be avoided becaouse of blast hazards caused by arc flash.
TRUSTED_SUSTAINABILITY
Specifications
Size Universal
Material Arc-rated Cotton, Nylon
Color Navy
Arc Flash Rating 12 cal/cm2
Certifications/Compliance ASTM, F2178, NFPA 70E™, ASTM F1506
Quantity 1 Ea.
Length (English) 18 in.
Length (Metric) 45.72 cm
Includes Universal hard hat bracket