Learn More
Honeywell Salisbury™ Pro-Wear™ Arc Flash Protection Jacket Kits
Catalog No. 19138164C Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-138-164C Large
19-138-164E 2X-Large
19-138-164B Medium
19-138-164F 3X-Large
19-138-164A Small
19-138-164D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-138-164C Supplier Honeywell Salisbury™ Supplier No. SK11LSPL
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
11cal/cm2 Flash Kit without gloves
Kits contain an arc flash jacket with hood, an AS1000HAT, SKBAG and safety glasses.
  • ATPV rating: 12cal/cm2
  • Jackets are made from arc flash resistant, Indura Ultra Soft™
  • Sewn with Nomex™ thread
  • Jacket length: 32 in. long
  • Nomex wristlets; hook and pile front closure
Order Info
Insulating rubber gloves are not included with these kits.
Certifications
Hazard Risk Category 2. CE marked, CE 0120 certified.
Compliance
Kits meet NFPA 70E-2009 and ASTM™ F1506 standards.
TRUSTED_SUSTAINABILITY
Specifications
Arc Flash Rating 11 cal/cm²
Description Coat, bib overalls, Pro-Hood™, hard hat, Salisbury storage bag and safety Glasses
Certifications/Compliance NFPA 70E™ 2009, CE 0120
Size Large