Learn More
Honeywell Salisbury™ Pro-Wear™ Arc Flash Protection Jacket Kits
Shop All WH Salisbury & Co. Products
Description
Kits contain an arc flash jacket with hood, an AS1000HAT, SKBAG and safety glasses.
- ATPV rating: 12cal/cm2
- Jackets are made from arc flash resistant, Indura Ultra Soft™
- Sewn with Nomex™ thread
- Jacket length: 32 in. long
- Nomex wristlets; hook and pile front closure
Order Info
Insulating rubber gloves are not included with these kits.
Certifications
Hazard Risk Category 2. CE marked, CE 0120 certified.
Compliance
Kits meet NFPA 70E-2009 and ASTM™ F1506 standards.
Specifications
Specifications
| Arc Flash Rating | 11 cal/cm² |
| Description | Coat, bib overalls, Pro-Hood™, hard hat, Salisbury storage bag and safety Glasses |
| Size | 3X-Large |
| Certifications/Compliance | NFPA 70E™ 2009, CE 0120 |