Learn More
Honeywell Salisbury™ PRO-WEAR™ Arc Flash Protective Coveralls
Catalog No. 19138121A Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-138-121A Small
19-138-121C Large
19-138-121B Medium
19-138-121F 3X-Large
19-138-121D X-Large
19-138-121E 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-138-121A Supplier Honeywell Salisbury™ Supplier No. ACCA11BLS
This item is not returnable. View return policy
11 cal/cm2 ATPV rating
  • Made from navy blue arc flash resistant Indura™ Ultra Soft™ fabric and sewn with Nomex™ thread
  • Full cut with set in sleeves
  • FR hook and pile front closure
  • 30 in. inseam
  • Expansion back
  • Hook and pile cuff opening
  • 2-ply Nomex wristlets
Compliance
Meet ASTM F1506 and NFPA 70E standards.
TRUSTED_SUSTAINABILITY
Specifications
Size Small
Material Indura™ Ultra soft™ Fabric
Color Navy
Arc Flash Rating 12 Calories/cm2
Certifications/Compliance ASTM F1506, NFPA 70E™, CE 0120
Size (English) Inseam 30 in.
Size (Metric) Inseam 76.2 cm