Learn More
Honeywell Salisbury™ PRO-WEAR PPE Arc Flash Kit
Catalog No. 19139222 Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Size: 2X-large
Catalog No.
19-139-222 Size: 2X-large
1 options
This item is not returnable. View return policy
Catalog No. 19-139-222 Supplier Honeywell Salisbury™ Supplier No. SK402XLBP
This item is not returnable. View return policy
Specifications
Arc Flash Rating 40 cal/cm²
Neck Style Hooded
Size 2X-Large
Certifications/Compliance ASTM F1506, NFPA 70E™, CE 0120
Description Coat and bib overalls, hood, hard hat, Salisbury back pack in place of the storage bag and safety Glasses
For Use With (Application) Personal protection