Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Salisbury™ Roll Blankets

Catalog No. 19138468 Shop All WH Salisbury & Co. Products
Encompass
Change view
Click to view available options
Material:
EPDM
Classification:
Class 0
Class 00
Class 1
Color:
Maroon
Orange, Yellow
Yellow
Voltage:
500 V
1000 V
7500 V
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Classification Color Voltage
19-138-468 EPDM Class 00 Maroon 500 V
19-138-469 EPDM Class 0 Yellow 1000 V
19-138-470 EPDM Class 1 Orange, Yellow 7500 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-138-468 Supplier Honeywell Salisbury™ Supplier No. RLB00
Only null left
Add to Cart
Add to Cart

Protection from low-voltage electrical hazards

  • Made from a high-strength fabric reinforced Type II rubber
  • All classes of blankets come in 3 x 30 ft. rolls that can be easily custom cut to fit each application
  • Self extinguishing and highly resistant to punctures, tears, oil and ozone
  • Unique colors make each class easy to identify and highly visible in the workplace
  • Class I clear PVC blanket allows complete visibility and provides the necessary insulating properties to meet ASTM F1742 standards
  • Blanket clamp pins, made of fiberglass reinforced nylon with molded rubber tips, can be used to hold blankets in place
TRUSTED_SUSTAINABILITY

Specifications

Color Maroon
Material EPDM
Certifications/Compliance ASTM F2320
Classification Class 00
Dimensions (L x W) 360 x 36 in. (914 x 91.44 cm)
For Use With (Application) Protection from low voltage electrical hazards, easily cut to fit
Length (English) 360 in.
Length (Metric) 914 cm
Voltage 500 V
Weight (English) 20 lb.
Weight (Metric) 9 kg
Width (English) 36 in.
Width (Metric) 91.44 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.