Learn More
Honeywell Salisbury™ Safety Kit
Catalog No. 19139220 Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Arc Flash Rating:
12 cal/cm²
42 cal/cm²
8 cal/cm²
Certifications/Compliance:
ASTM F1506, NFPA 70E™, CE 0120
NFPA 70E™ 2009, CE 0120
Size:
Large
X-Large
2X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Arc Flash Rating Certifications/Compliance Size
19-139-220 42 cal/cm² ASTM F1506, NFPA 70E™, CE 0120 Large
19-139-213 8 cal/cm² NFPA 70E™ 2009, CE 0120 Large
19-139-214 8 cal/cm² NFPA 70E™ 2009, CE 0120 X-Large
19-139-217 12 cal/cm² NFPA 70E™ 2009, CE 0120 Large
19-139-215 8 cal/cm² NFPA 70E™ 2009, CE 0120 2X-Large
19-139-218 12 cal/cm² NFPA 70E™ 2009, CE 0120 X-Large
19-139-219 12 cal/cm² NFPA 70E™ 2009, CE 0120 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-139-220 Supplier Honeywell Salisbury™ Supplier No. SK40LBP
This item is not returnable. View return policy
Specifications
Arc Flash Rating 42 cal/cm²
Description AS1200 hat, Salisbury back pack, hard hat and safety Glasses
Size Large
Certifications/Compliance ASTM F1506, NFPA 70E™, CE 0120