Learn More
Drg International Inc Saliva 17Oh Progesterone Elisa
Supplier:  Drg International Inc SLV3140
Encompass_Preferred
An enzyme immunoassay for the quantitative in vitro diagnostic measurement of active free 17 hydroxyprogesterone in saliva. Measurements of 17-hydroxyprogesterone are used as an aid in the diagnosis of various disorders of the adrenal glands or the ovaries, and as an aid in the diagnosis of late onset of 21-hydroxylase deficiency, a common cause of Congenital Adrenal Hyperplasia. This test is not intended for newborn screening. The DRG Salivary 17Alpha-OHP ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a polyclonal antibody (rabbit) directed towards an antigenic site on the 17Alpha-OHP molecule. Endogenous 17Alpha-OHP of a patient sample competes with a 17Alpha-OHP-horseradish peroxidase conjugate for binding to the coated antibody.
Catalog No. 50-194-7639