Learn More
NEST SCIENTIFIC USA SALIVA COLLECTION KIT 10ML
Supplier:  NEST SCIENTIFIC USA 203112
Encompass_Preferred
Saliva Collection Kit with ITM, each kit Includes a 10 mL Vial with a funnel, a separate 10 mL Vial filled with 2.0 mL ITM, Sterile, 100 kits/case. Funnel and tube made from USP grade VI polypropylene. Molded in ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. Produced in a cGMP (21 CFR part 820/821), ISO 13485 and ISO 9001 facility.
Catalog No. NC2958277