Learn More
G-Biosciences Saliva Collection Kit
SCI_ED
Catalog No. S04211
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
S04211 10/Pk.
1 options
Catalog No. S04211 Supplier G-Biosciences Supplier No. 7862186
Add to Cart
Add to Cart
Saliva collection kit is a non-invasive sample collection device for stabilization, transport and storage of human saliva.
Saliva collection kit is a non-invasive sample collection device for stabilization, transport and storage of human saliva. It is suitable for at-home self collection. It is designed to easily capture and store saliva samples deposited directly from patients.
This kit includes:
  • Large funnel for easy sample depositing; disconnects for disposal after use.
  • 6 mL standardized polypropylene tube with cap, compatible with automation rack.
  • Optimized concentration of Proteinase-K to preserve integrity of DNA/RNA.
Specifications
Product Type Saliva Collection Kit
Quantity 10/Pk.