Learn More
G-Biosciences Saliva Collection Tubes
SCI_ED
Catalog No. S04210
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
S04210 10/Pk.
1 options
Catalog No. S04210 Supplier G-Biosciences Supplier No. 7862185
Add to Cart
Add to Cart
Saliva collection tube is a non-invasive sample collection device for stabilization, transport and storage of human saliva.
Saliva collection tube is a non-invasive sample collection device for stabilization, transport and storage of human saliva. It is suitable for at-home self collection.
Specifications
Product Type Saliva Collection Device
Quantity 10/Pk.