Learn More
Pickering Laboratories Inc Artificial Saliva, DIN 53160, Not Stabilized, 2-part mix, each (200 mL/bottle)
SDP
Catalog No. 501354828
Encompass_Preferred
Artificial saliva is prepared according to specifications of DIN 53160-1:200, SN/T 1058.1-2013 and GB/T 18886-2019 official methods. The formulation consists of two parts that are mixed right before use. This minimizes changes in the solution during storage and allows for a longer shelf life. The solution should be kept frozen for long-term storage. The pH of the solution after mixing is 6.8. A made-to-order pre-mixed formulation is available for customers who intend to use the artificial saliva within one month
Catalog No. 50-135-4828 Supplier Pickering Laboratories Inc Supplier No. 17000303
Add to Cart
Add to Cart
Artificial saliva is prepared according to specifications of DIN 53160-1:200, SN/T 1058.1-2013 and GB/T 18886-2019 official methods. The formulation consists of two parts that are mixed right before use. This minimizes changes in the solution during storage and allows for a longer shelf life. The solution should be kept frozen for long-term storage. The pH of the solution after mixing is 6.8. A made-to-order pre-mixed formulation is available for customers who intend to use the artificial saliva within one month