Learn More
Norgen Biotek Corp Saliva DNA Collection and Preservation Devices, 50 Pack
Catalog No. NC1385022
Encompass_Preferred
Norgen's Saliva DNA Collection and Preservation Devices are designed for 1) simple and non-invasive saliva collection and 2) preservation of DNA in saliva samples at ambient temperature. Each of the 50 Saliva DNA Collection and Preservation Devices consists of 3 components: (1) Saliva Collection Funnel and Collection Tube, (2) Collection Tube Cap, and (3) Norgen's Saliva DNA Preservative contained within a sealed squeezable ampoule. Saliva samples are collected by spitting inside the Collection Funnel which has been assembled with the Collection Tube. After collecting the required volume of saliva the Collection Funnel is removed and the contents of the Preservative Ampoule are then added and mixed with the collected saliva. The Saliva Collection Tube is subsequently sent to the laboratory for DNA isolation and analysis.
Catalog No. NC1385022 Supplier Norgen Biotek Corp Supplier No. RU49000
Norgen's Saliva DNA Collection and Preservation Devices are designed for 1) simple and non-invasive saliva collection and 2) preservation of DNA in saliva samples at ambient temperature. Each of the 50 Saliva DNA Collection and Preservation Devices consists of 3 components: (1) Saliva Collection Funnel and Collection Tube, (2) Collection Tube Cap, and (3) Norgen's Saliva DNA Preservative contained within a sealed squeezable ampoule. Saliva samples are collected by spitting inside the Collection Funnel which has been assembled with the Collection Tube. After collecting the required volume of saliva the Collection Funnel is removed and the contents of the Preservative Ampoule are then added and mixed with the collected saliva. The Saliva Collection Tube is subsequently sent to the laboratory for DNA isolation and analysis.