Learn More
Boca Scientific Inc GeneFiX Saliva-Prep 2 Isolation Kit, 50 Reactions
Catalog No. 501685620
Encompass
Isohelix GeneFiX Saliva-Prep 2 Isolation kit is specifically designed for use with saliva collection samples. It uses a new formulated precipitation method to quickly and easily isolate the DNA, up to volumes of 2ml from the GeneFix Saliva Collectors. The GeneFiX Saliva-Prep 2 kit does not use solvents, columns or filtration. The reduced number of protocol steps makes it extremely easy-to-use when a simple yet efficient isolation method is required.
Catalog No. 50-168-5620 Supplier Boca Scientific Inc Supplier No. GSPN50
Isohelix GeneFiX Saliva-Prep 2 Isolation kit is specifically designed for use with saliva collection samples. It uses a new formulated precipitation method to quickly and easily isolate the DNA, up to volumes of 2ml from the GeneFix Saliva Collectors. The GeneFiX Saliva-Prep 2 kit does not use solvents, columns or filtration. The reduced number of protocol steps makes it extremely easy-to-use when a simple yet efficient isolation method is required.