Learn More
Pickering Laboratories Inc Artificial Saliva, Pharmaceutical Research, Not Stabilized, each (200 mL/bottle)
SDP
Catalog No. 501354829
Encompass_Preferred
Artificial Saliva is formulated according to literature for pharmaceutical research such as studies of drug dissolution and drug delivery through oral mucosa. This is a ready to use formulation that should be stored refrigerated. The pH of the solution is 6.8
Catalog No. 50-135-4829 Supplier Pickering Laboratories Inc Supplier No. 17000304
Artificial Saliva is formulated according to literature for pharmaceutical research such as studies of drug dissolution and drug delivery through oral mucosa. This is a ready to use formulation that should be stored refrigerated. The pH of the solution is 6.8