Learn More
Norgen Biotek Corp SALIVA RNA COLLECT & PRESERVE
Catalog No. NC1810680
Encompass_Preferred
Norgen's Saliva RNA Collection and Preservation Devices are designed for 1) simple and non-invasive saliva collection and 2) preservation of RNA in saliva samples at ambient temperature. Each of the 50 Saliva RNA Collection and Preservation Devices consists of 3 components: (1) Saliva Collection Funnel and Collection Tube, (2) Collection Tube Cap, and (3) Norgen's Saliva RNA Preservative contained within a sealed squeezable ampoule. After collecting the required volume of saliva the Collection Funnel is removed and the contents of the Preservative Ampoule are then added and mixed with the collected saliva. RNA can be isolated from the preserved saliva samples using Norgen's Saliva/Swab RNA Purification Kits (Cat# 69100/69300). Each of Norgen's Collection Tubes is labeled with a unique serial number for secure and anonymous tracking of the sample. Saliva RNA in preserved samples is stable for up to 2 months at room temperature.
Catalog No. NC1810680 Supplier Norgen Biotek Corp Supplier No. RU53800
Norgen's Saliva RNA Collection and Preservation Devices are designed for 1) simple and non-invasive saliva collection and 2) preservation of RNA in saliva samples at ambient temperature. Each of the 50 Saliva RNA Collection and Preservation Devices consists of 3 components: (1) Saliva Collection Funnel and Collection Tube, (2) Collection Tube Cap, and (3) Norgen's Saliva RNA Preservative contained within a sealed squeezable ampoule. After collecting the required volume of saliva the Collection Funnel is removed and the contents of the Preservative Ampoule are then added and mixed with the collected saliva. RNA can be isolated from the preserved saliva samples using Norgen's Saliva/Swab RNA Purification Kits (Cat# 69100/69300). Each of Norgen's Collection Tubes is labeled with a unique serial number for secure and anonymous tracking of the sample. Saliva RNA in preserved samples is stable for up to 2 months at room temperature.