Learn More
Drg International Inc Salivary Dhea-S Elisa Ruo
Supplier:  Drg International Inc SLV4409R
Encompass_Preferred
A competitive immunoenzymatic colorimetric method for quantitative determination of DHEA-S concentration in saliva. This product is for Research Use Only. The DHEA-S (antigen) in the sample competes with the antigenic DHEA-S conjugated with horseradish peroxidase (HRP) for binding to the limited number of antibodies anti DHEA-S coated on the microplate (solid phase).
Catalog No. 50-194-7641