Learn More
Drg International Inc Salivary Progesterone Hs Elisa
Supplier:  Drg International Inc SLV5911
Encompass_Preferred
An enzyme immunoassay for the in vitro diagnostic quantitative measurement of active free progesterone (a female hormone) in saliva. Measurements obtained by this device may be used in the diagnosis and treatment of disorders of the ovaries or placenta and can be used as an aid for prediction of ovulation. Progesterone (4-pregnene-3, 20-dione) is a C21 steroid hormone containing a keto-group (at C-3) and a double bond between C-4 and C-5. The Progesterone level in saliva represents the concentration of the active free Progesterone. The DRG Salivary Progesterone ELISA kit is based on the competition principle and the microplate separation. An unknown amount of Progesterone present in the sample and a fixed amount of Progesterone conjugated with horseradish peroxidase compete for the binding sites of a rabbit polyclonal Progesterone -antiserum coated onto the wells.
Catalog No. 50-194-7636