Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP10146MA20UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: Flag . Host: Mammal. Species Reactivity: Human. Protein Expression: A DNA sequence encoding the Homo sapiens (Human) Sal-like protein 2, was expressed in the hosts and tags indicated. AA Sequence: MSRRKQRKPQ QLISDCEGPS ASENGDASEE DHPQVCAKCC AQFTDPTEFL AHQNACSTDP PVMVIIGGQE NPNNSSASSE PRPEGHNNPQ VMDTEHSNPP DSGSSVPTDP TWGPERRGEE SPGHFLVAAT GTAAGGGGGL ILASPKLGAT PLPPESTPAP PPPPPPPPPP GVGSGHLNIP LILEELRVLQ QRQIHQMQ. Buffer: Tris-based buffer, 50% glycerol. Purity: Greater than 90% as determined by SDS-PAGE. Endotoxin: Not determined. Gene Symbol: SALL2. Gene ID: 6297. Uniprot: Q9Y467.
Catalog No. 50-188-5504