Learn More
Diagnostic Biosystems SALL4 SALL4 (6E3)
Catalog No. 502407321
Encompass_Preferred
Sal-like protein 4 (SALL4) is a zinc-finger transcription factor that serves as a master regulator of embryonic pluripotency and is involved in processes associated with stem cell activities. SALL4 expression in germ cells makes it a useful marker for germ cell tumors such as seminoma, embryonal carcinoma, yolk sac tumors and teratomas. SALL4 expression is also seen in the spermatogonia of normal testis.
Catalog No. 50-240-7321 Supplier Diagnostic Biosystems Supplier No. PDM591
Sal-like protein 4 (SALL4) is a zinc-finger transcription factor that serves as a master regulator of embryonic pluripotency and is involved in processes associated with stem cell activities. SALL4 expression in germ cells makes it a useful marker for germ cell tumors such as seminoma, embryonal carcinoma, yolk sac tumors and teratomas. SALL4 expression is also seen in the spermatogonia of normal testis.