Learn More
Cederlane SALM.O GROUP ANTISERA,O:18 3ML
SDP
Catalog No. NC0410910
Encompass
(Product: O:18),(Reacts with factor:K ),(No. of tests: 150),(SSI� SALMONELLA O GROUP ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial
Catalog No. NC0410910 Supplier Cederlane Supplier No. 40232(SS)
(Product: O:18),(Reacts with factor:K ),(No. of tests: 150),(SSI� SALMONELLA O GROUP ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial