Learn More
Medchemexpress LLC HY-14302 10mg Medchemexpress, Salmeterol CAS:89365-50-4 Purity:>98%
SDP
Catalog No. 502081587
Encompass_Preferred
Medchemexpress, HY-14302 10mg Salmeterol CAS:89365-50-4 Salmeterol (GR33343X) is a potent and selective human β2 adrenoceptor agonist. Salmeterol shows potent stimulation of cAMP accumulation in CHO cells expressing human β2, β1 and β3 adrenoceptors with pEC50s of 9.6, 6.1, and 5.9, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1587 Supplier Medchemexpress LLC Supplier No. HY1430210MG
Medchemexpress, HY-14302 10mg Salmeterol CAS:89365-50-4 Salmeterol (GR33343X) is a potent and selective human β2 adrenoceptor agonist. Salmeterol shows potent stimulation of cAMP accumulation in CHO cells expressing human β2, β1 and β3 adrenoceptors with pEC50s of 9.6, 6.1, and 5.9, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.