Learn More
Selleck Chemical LLC Salmeterol 5mg 89365-50-4 Astmerole, GR-33343X, SN408D
SDP
Catalog No. 501948355
Encompass_Preferred
Salmeterol (Astmerole, GR-33343X, SN408D) is a long-acting beta2-adrenergic receptor agonist with Ki value of 1.5 nM for WT ?2AR and shows very high selectivity for the WT ?2AR(?1Ki/?2Ki ratio of approximately 1500). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8355 Supplier Selleck Chemical LLC Supplier No. S55275MG
Salmeterol (Astmerole, GR-33343X, SN408D) is a long-acting beta2-adrenergic receptor agonist with Ki value of 1.5 nM for WT ?2AR and shows very high selectivity for the WT ?2AR(?1Ki/?2Ki ratio of approximately 1500). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.