Learn More
Cayman Chemical Salmeterol-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 497063-94-2, 1 mg
SDP
Catalog No. 502933612
Encompass_Preferred
An internal standard for the quantification of salmeterol by GC- or LC-MS
Catalog No. 50-293-3612 Supplier Cayman Chemical Supplier No. 264491 MG
An internal standard for the quantification of salmeterol by GC- or LC-MS