Learn More
Medchemexpress LLC HY-17453 10mg Medchemexpress, Salmeterol (xinafoate) CAS:94749-08-3 Purity:>98%
SDP
Catalog No. 502081956
Encompass_Preferred
Medchemexpress, HY-17453 10mg Salmeterol (xinafoate) CAS:94749-08-3 Salmeterol (GR 33343X) xinafoate is a potent and selective human β2 adrenoceptor agonist. Salmeterol shows potent stimulation of cAMP accumulation in CHO cells expressing human β2, β1 and β3 adrenoceptors with pEC50s of 9.6, 6.1, and 5.9, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1956 Supplier Medchemexpress LLC Supplier No. HY1745310MG
Medchemexpress, HY-17453 10mg Salmeterol (xinafoate) CAS:94749-08-3 Salmeterol (GR 33343X) xinafoate is a potent and selective human β2 adrenoceptor agonist. Salmeterol shows potent stimulation of cAMP accumulation in CHO cells expressing human β2, β1 and β3 adrenoceptors with pEC50s of 9.6, 6.1, and 5.9, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.