Learn More
TARGETMOL CHEMICALS INC SALMETEROL XINAFOATE 25MG
SDP
Catalog No. 502579316
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Salmeterol Xinafoate (GR 33343X xinafoate) is a long-acting (beta)2-adrenergic receptor agonist with anti-inflammatory effects used in the treatment of asthma symptoms and chronic obstructive pulmonary disease (COPD) symptoms. purity: 98%
Catalog No. 50-257-9316 Supplier TARGETMOL CHEMICALS INC Supplier No. T665325MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Salmeterol Xinafoate (GR 33343X xinafoate) is a long-acting (beta)2-adrenergic receptor agonist with anti-inflammatory effects used in the treatment of asthma symptoms and chronic obstructive pulmonary disease (COPD) symptoms. purity: 98%