Learn More
Cayman Chemical SalmeterolxInafoate 50mg
SDP
Catalog No. 502969220
Encompass_Preferred
A long-acting β2-AR agonist (EC50s = 0.79, 63.1, and 9.4 nM for β2-, β1-, and β3-ARs, respectively); inhibits electrically-stimulated contraction of isolated guinea pig trachea strips (EC50 = 2.51 nM); inhibits histamine-induced bronchoconstriction in guinea pigs via aerosol administration at 0.12-12 mM
Catalog No. 50-296-9220 Supplier Cayman Chemical Supplier No. 1600950 MG
A long-acting β2-AR agonist (EC50s = 0.79, 63.1, and 9.4 nM for β2-, β1-, and β3-ARs, respectively); inhibits electrically-stimulated contraction of isolated guinea pig trachea strips (EC50 = 2.51 nM); inhibits histamine-induced bronchoconstriction in guinea pigs via aerosol administration at 0.12-12 mM