Learn More
Abcam Salmon Sperm ssDNA, 5MG
Catalog No. 502586972
Encompass_Preferred
This product is a convenient, ready-to-use solution of salmon sperm DNA especially prepared for use in the preparation of pre-hybridization and hybridization solutions and as a DNA carrier in yeast transformation protocols and other related methods.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-6972 Supplier Abcam Supplier No. AB2292785MG
This product is a convenient, ready-to-use solution of salmon sperm DNA especially prepared for use in the preparation of pre-hybridization and hybridization solutions and as a DNA carrier in yeast transformation protocols and other related methods.
The product is subject to the following: Abcam Restricted Use Statement