Learn More
Kingfisher Biotech Inc Bovine CCL2 (MCP-1) ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1543AS003
Encompass_Preferred
The Bovine CCL2 (MCP-1) ELISA contains capture antibody, protein standard, and detection antibody for development of a Bovine CCL2 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The Bovine CCL2 ELISA can also be used to measure Wild Yak CCL2. Alternate names include CCL2, GDCF-2, HC11, HSMCR30, MCAF, MCP-1, MCP1, SCYA2, SMC-CF, C-C motif chemokine ligand 2. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5517