Learn More
Cederlane SALMONELLA 0:23 150TST
SDP
Catalog No. NC1381877
Encompass
(Product: O:23),(Reacts with factor:G),(No. of tests: 150),(SSI® SALMONELLA O FACTOR ANTISERA FOR SLIDE AGGLUTINATION)
Catalog No. NC1381877 Supplier Cederlane Supplier No. 40283(SS)
(Product: O:23),(Reacts with factor:G),(No. of tests: 150),(SSI® SALMONELLA O FACTOR ANTISERA FOR SLIDE AGGLUTINATION)