Learn More
Cederlane SALMONELLA ANTISERA OMA 3ML
SDP
Catalog No. NC0486846
Encompass
(Product: OMA),(Reacts with factor:A, B, D, E, L 1, 2, 3, 4, 5, 9, 10, 12, 15, 19, 21, 46),(No. of tests: 150),(SSI� SALMONELLA O GROUP POOL ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial
Catalog No. NC0486846 Supplier Cederlane Supplier No. 40212(SS)
(Product: OMA),(Reacts with factor:A, B, D, E, L 1, 2, 3, 4, 5, 9, 10, 12, 15, 19, 21, 46),(No. of tests: 150),(SSI� SALMONELLA O GROUP POOL ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial