Learn More
ATCC Salmonella enterica; serovar Typhimurium; Quantitative DNA
Catalog No. 502383626
Encompass
Quantitative genomic DNA from Salmonella enterica subsp. enterica serovar Typhimurium strain LT2 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load and for other molecular biology applications.
Catalog No. 50-238-3626 Supplier ATCC Supplier No. 700720DQ
Quantitative genomic DNA from Salmonella enterica subsp. enterica serovar Typhimurium strain LT2 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load and for other molecular biology applications.