Learn More
ATCC Nitrosomonas stercoris;Strain: KYUHI-S
Catalog No. 502380167
Encompass
Type strain. A chemoautotrophic ammonia-oxidizing bacterium tolerant of high ammonium.
Catalog No. 50-238-0167 Supplier ATCC Supplier No. BAA2718
Type strain. A chemoautotrophic ammonia-oxidizing bacterium tolerant of high ammonium.